Anti-C-ABL Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24479
89361-456EA 727.02 CAD
89361-456
Anti-C-ABL Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to c - Abl ( phospho Y245 )

WB: Use at a concentration of 0.1 - 1.0 ?g/ml. Detects a band of approximately 146 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: C-ABL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR