c-Abl is a 140-150 kDa non-receptor protein tyrosine kinase whose precise functions are not known, but roles for Abl in growth factor and integrin signaling, cell cycle regulation, cytoskeletal reorganization, neurogenesis, and responses to DNA damage and oxidative stress have been suggested. c-Abl kinase activity is increased in vivo by diverse physiological stimuli including ionizing radiation, entry into S phase, integrin activation, and platelet-derived growth factor (PDGF) stimulation. c-Abl contains various protein binding domains that appear to enable it to regulate the functions of many proteins by forming complexes, most notably three isoforms of the oncogenic protein Bcr/Abl. c-Abl becomes fully activated by sequential phosphorylation of tyrosines 412 and 245.
Type: Primary
Antigen: C-ABL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human