Anti-CAD Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX28406
89352-482EA 830.57 CAD
89352-482
Anti-CAD Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to CAD

Western blot at 1:500 to 1:1000 dilution.

Type: Primary
Antigen: CAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR