Anti-CAF1A Mouse Monoclonal Antibody [clone: SS 53]

GTX30248
89367-662EA 630.26 CAD
89367-662
Anti-CAF1A Mouse Monoclonal Antibody [clone: SS 53]
Chromatin assembly factor 1 (CAF-1) is a member of the histone chaperone family. It is required for inheritance of epigenetically determined chromosomal states, and in vitro, it promotes the assembly of chromatin during DNA replication and DNA repair. CAF-1 is a complex of three subunits; p150, p60, and p48. This complex is localized to the nucleus of human cells. Both the composition and the biochemical activity of CAF-1 are conserved among human, yeast, and Drosophila.

Type: Primary
Antigen: CAF1A
Clonality: Monoclonal
Clone: SS 53
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR