Anti-CAFFEINE Mouse Monoclonal Antibody

Supplier: GeneTex

GTX15221
89360-114EA 890.5 CAD
89360-114
Anti-CAFFEINE Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CAFFEINE
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR