Anti-CSP1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX100245
89348-312EA 673.08 CAD
89348-312
Anti-CSP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Calcipressin 1 (regulator of calcineurin 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CSP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR