Anti-CALCA Rabbit Monoclonal Antibody [clone: SP17]

Supplier: Genetex

GTX79426
89353-854EA 711.84 CAD
89353-854
Anti-CALCA Rabbit Monoclonal Antibody [clone: SP17]
Antibodies
Monoclonal Rabbit Antibody to Human Calcitonin (ready-to-use)

We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining.

Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR