Anti-HP Rabbit Polyclonal Antibody

GTX77233
89353-564EA 407.48 CAD
89353-564
Anti-HP Rabbit Polyclonal Antibody
Affinity Purified Rabbit anti - Rat Haptoglobin

For ELISA: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: HP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR