Anti-APOC1 Goat Polyclonal Antibody

Supplier: GeneTex

GTX27617
89362-616EA 467.76 CAD
89362-616
Anti-APOC1 Goat Polyclonal Antibody
Antibodies
Goat Polyclonal antibody to Apolipoprotein C-I

Recommended Starting Dilutions:WB: Use at 1:5,000 - 1:10,000ELISA: Use at 1:5,000 - 1:10,000IHC: Use at 1:50 - 1:200IP: Use at 1:100

Type: Primary
Antigen: APOC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR