Anti-CRP55 Sheep Polyclonal Antibody

Supplier: GeneTex
GTX16144
89360-184EA 789.92 CAD
89360-184
Anti-CRP55 Sheep Polyclonal Antibody
Antibodies
Calreticulin is an abundant highly conserved calcium binding protein found in the lumen of the endoplasmic reticulum of eukaryotic cells. This lumen protein can be used to measure the intactness of membrane preparations.

  • For IF: Use at a dilution of 1:50. For WB: Use at a dilution of 1:1,000 - 1:5,000. Predicted molecular weight: 48 kDa. Not tested in other applications. Could be used as a marker for subcellular fractionation of cells. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CRP55
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR