Anti-CD14 Mouse Monoclonal Antibody [clone: MEM-18]

Supplier: GeneTex

GTX75931
89353-384EA 624.21 CAD
89353-384
Anti-CD14 Mouse Monoclonal Antibody [clone: MEM-18]
Antibodies
Mouse Antibody to Human CD14:FITC

Flow Cytometry: Use 20?l of the suggested working dilution to label 10^6 cells in 100?l. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR