Anti-CD14 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX112890
89357-216EA 623.96 CAD
89357-216
Anti-CD14 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CD14 (CD14 molecule)

Suggested starting dilutions are as follows: WB: 1:500-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CD14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR