Anti-TM Mouse Monoclonal Antibody [clone: QBEND/40]

Supplier: GeneTex

GTX76378
89366-074EA 330.79 CAD
89366-074
Anti-TM Mouse Monoclonal Antibody [clone: QBEND/40]
Antibodies
Mouse monoclonal antibody [QBEND/40] to Thrombomodulin

Recommended Starting Dilutions: Immunohistology - Frozen: 1/10 - 1/100 Immunohistology - Paraffin: 1/10 - 1/50 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: TM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR