Anti-CD44 Rabbit Polyclonal Antibody

GTX100499
89348-504EA 472.59 CAD
89348-504
Anti-CD44 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to CD44 (CD44 molecule (Indian blood group))

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CD44
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR