Anti-IgG Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX27156
89362-462EA 193.83 CAD
89362-462
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
FITC-conjugated Rabbit anti-Human IgG polyclonal antibody, pre-adsorbed

Recommended Starting Dilutions:FACS: Use at 1:2,000-1:10,000. For IM: Use at 1:500-1:2,000.Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR