Anti-IL4 Rabbit Polyclonal Antibody

GTX29622
89367-642EA 472.59 CAD
89367-642
Anti-IL4 Rabbit Polyclonal Antibody
Rabbit polyclonal to IL4

Neutralization: 0.5 - 1 ?g/ml. WB: 0.5 - 1 ?g/ml. However, the optimal conditions should be determined individually.

Type: Primary
Antigen: IL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR