Anti-LEP Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX17629
89360-318EA 673.19 CAD
89360-318
Anti-LEP Rabbit Polyclonal Antibody
Antibodies
Leptin is an anorexigenic hormone produced in adipocytes which has wide ranging functions beyond satiety. Many of leptin's affects are mediated centrally (esp.the hypothalamus).

ELISA: 1/1000 - 1/4000. WB: 1/1000 - 1/4000. Predicted molecular weight: 16 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR