Anti-CD68 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX101866
89349-318EA 623.96 CAD
89349-318
Anti-CD68 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CD68 (macrosialin)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CD68
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR