Anti-CHRM2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX71171
89365-198EA 677.37 CAD
89365-198
Anti-CHRM2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to mAChR M2

Immunohistochemistry with formalin fixed paraffin embedded tissues. Not tested for other uses. IHC-P: Use at a dilution of 1/100 - 1/200.

Type: Primary
Antigen: CHRM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR