Anti-CHRM5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX70523
89365-028EA 677.37 CAD
89365-028
Anti-CHRM5 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to mAChR M5

IHC-P: Use at a dilution of 1/100 - 1/200.

Type: Primary
Antigen: CHRM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR