Anti-DR6 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX74386
89365-444EA 667.28 CAD
89365-444
Anti-DR6 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to DR6

Recommended Starting Dilutions: Western Blotting: Max Dilution 1/500 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: DR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR