Anti-DR6 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX28417
89352-486EA 781.14 CAD
89352-486
Anti-DR6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to DR6

Western blot at 1:500 to 1:2000 dilution.

Type: Primary
Antigen: DR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR