Anti-ECS Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX30528
89352-688EA 699.51 CAD
89352-688
Anti-ECS Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ECS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR