Anti-MAX Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX11984
89359-080EA 582.61 CAD
89359-080
Anti-MAX Rabbit Polyclonal Antibody
Antibodies
A basic helix-loop-helix, leucine zipper protein that forms heterodimers with MYC and MAD - regulating gene expression.

WB: Use at a dilution of 1/500 - 1/1000. Detects a band of approximately 18 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MAX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR