Anti-MCP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX29779
89362-846EA 709.4 CAD
89362-846
Anti-MCP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to MCP1

For WB: Use at a dilution of 1:1,500 - 1:2,500. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR