Anti-IgG2a Goat Polyclonal Antibody

Supplier: Genetex

GTX29163
89352-586EA 291.54 CAD
89352-586
Anti-IgG2a Goat Polyclonal Antibody
Antibodies
This secondary antibody specifically recognizes Mouse IgG2a. It does not react with any other isotypes.

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR