Anti-MCAM Mouse Monoclonal Antibody [clone: OJ79c]

Supplier: GeneTex

GTX75873
89367-930EA 673.44 CAD
89367-930
Anti-MCAM Mouse Monoclonal Antibody [clone: OJ79c]
Antibodies
Mouse monoclonal antibody [OJ79c] to CD146

Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: MCAM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR