Anti-MYO5A Rabbit Polyclonal Antibody

GTX11092
89356-496EA 456.78 CAD
89356-496
Anti-MYO5A Rabbit Polyclonal Antibody
Rabbit polyclonal to MYO5A

IHC-P: Use at a dilution of 1/200. WB: Use at a dilution of 1/500. Detects a band of approximately 190 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MYO5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR