Anti-P57KIP2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX24058
89352-092EA 678.26 CAD
89352-092
Anti-P57KIP2 Rabbit Polyclonal Antibody
Rabbit polyclonal to p57 Kip2

Immunohistochemistry: use at a dilution of 1/25 - 1/50 for 30mins at RT (ABC method). Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. For formalin fixed paraffin embedded sections, perform heat mediated antigen retrieval using 10mM citrate buffer pH 6.0 before commencing with the IHC staining protocol.

Type: Primary
Antigen: P57KIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR