Anti-FLIP Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX59693
89363-732EA 767.76 CAD
89363-732
Anti-FLIP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to c-FLIP

Western blot analysis (0.5-4 µg/ml) and immunofluorescence. However, the optimal conditions should be determined individually. The immunoaffinity-purified antibody detects 58 kDa FLIPL on SDS-PAGE immunoblots, in samples from human, mouse and rat origins. The antibody recognizes only the FLIPL form.

Type: Primary
Antigen: CFLAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR