Anti-FPR3 Rabbit Polyclonal Antibody

GTX71837
89365-328EA 548.88 CAD
89365-328
Anti-FPR3 Rabbit Polyclonal Antibody
FPRL2 is a Chemoattractant Receptor involved in the transduction of signals in myeloid cells. Although it is structurally homologous to FPRL1, human FPRL2 does not recognize the N-formylmethionyl peptides (FMLP) as ligands and is not expressed in neutrophils.

Type: Primary
Antigen: FPRL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR