Anti-GALR1 Rabbit Polyclonal Antibody

GTX71465
89365-266EA 633.57 CAD
89365-266
Anti-GALR1 Rabbit Polyclonal Antibody
Rabbit Polyclonal to Human Galanin Receptor GalR1

Immunohistochemistry with formalin fixed paraffin embedded tissues. Not tested for other uses. IHC-P: Use at a dilution of 1/100 - 1/200.

Type: Primary
Antigen: GALR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR