Anti-GPCR GPRC5D Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX78097
89366-500EA 701.09 CAD
89366-500
Anti-GPCR GPRC5D Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to G Protein-Coupled Receptor GPRC5D

Type: Primary
Antigen: GPRC5D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR