Anti-IgG Rabbit Polyclonal Antibody

Supplier: Genetex

GTX26772
89352-200EA 558.59 CAD
89352-200
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Immunoglobulin G (IgG) is one of the most abundant protein in serum with normal levels between 8-17 mg/ml in adult blood. IgG is important for our defense against microorganisms and the molecules are produced by B-lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions: to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR