Anti-HIST1H2BB Rabbit Polyclonal Antibody

GTX61374
89363-956EA 520.01 CAD
89363-956
Anti-HIST1H2BB Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Histone H2B

Recommended Starting Dilutions: For WB: Use at a dilution of 1:20,000 For IHC: Use at a dilution of 1:100 - 250 For ICC: Use at a dilution of 1:50 - 100 For IP: Use at a dilution of 1:50 Optimal working dilution for a specific application should be determined by the investigator.

Type: Primary
Antigen: HIST1H2BB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR