Anti-IgG Sheep Polyclonal Antibody

Supplier: GeneTex
GTX26870
89362-214EA 314.42 CAD
89362-214
Anti-IgG Sheep Polyclonal Antibody
Antibodies
HRP-conjugated Sheep anti-Human IgG polyclonal antibody

Recommended Starting Dilutions: For ELISA : 1:150,000 - 1:250,000. For WB : 1:1,000 - 1:10,000. For IHC : 1:500 - 1:2,500. Optimal titers for other applications should be determined by the researcher.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR