Anti-IgE Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX73360
89353-166EA 565.77 CAD
89353-166
Anti-IgE Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with the epsilon chain of human IgE. Traces of contaminating antibodies have been removed by solid phase absorption. Only the IgE arch appeared in crossed immunoelectrophoresis.

Type: Primary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR