Anti-IL2 Goat Polyclonal Antibody

GTX10752
89354-990EA 456.78 CAD
89354-990
Anti-IL2 Goat Polyclonal Antibody
Goat polyclonal to IL2

ELISA: Use at a concentration of 1 ?g/ml to detect 0.6 ng/well of recombinant human IL2. Neut: Use at a concentration of 0.8 - 8.0 ?g/ml. WB: Use at a concentration of 2 ?g/ml, this detects 5 ng/lane of recombinant human IL2 under reducing conditions and 20 ng/lane under non-reducing conditions. Predicted molecular weight: 19 kDa. Anti-Human IL2 is tested for its ability to neutralize the biological activity of recombinant, human IL2 on CTLL2 cells. The ND50 of the antibody is defined as the concentration of antibody resulting in a one-half maximal inhibition of bioactivity of recombinant, human IL2, when IL2 is present at a concentration just high enough to elicit a maximum response. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR