Anti-IL2 Rabbit Polyclonal Antibody

GTX74203
89365-418EA 522.15 CAD
89365-418
Anti-IL2 Rabbit Polyclonal Antibody
Recognizes natural and recombinant human IL-2.

Type: Primary
Antigen: IL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR