Anti-SOD2 Goat Polyclonal Antibody

Supplier: GeneTex
GTX11888
89358-928EA 922.29 CAD
89358-928
Anti-SOD2 Goat Polyclonal Antibody
Antibodies
Superoxide dismutase (SOD) is responsible for the elimination of cytotoxic active oxygen by catalyzing the dismutation of the superoxide radical to oxygen and hydrogen peroxide. There are three SOD isoenzymes in mammalian cells. They are: extracellular SOD (EC SOD), copper and zinc-containing SOD (Cu/Zn SOD) and manganese-containing SOD (Mn SOD). The Cu/Zn form contains Cu and Zn ions and exists as a 32 kDa dimer in the cytosol. Mn SOD is an 80 kDa tetramer that contains Mn ion and resides in the mitochondrial matrix. Mn SOD is a tumor necrosis factor (TNF)-inducible enzyme that protects cells from TNF-mediated apoptosis via superoxide anion detoxification and the subsequent regulation of apoptosis through cytochrome c release and the modulation of the redox state of the mitochondria. Mn SOD has also been shown to be a tumor suppressor in human breast cancer. Overexpression of this enzyme protects neurons from NMDA- and nitric oxide-induced neurotoxicity.

WB: Use at a dilution of 1/1000 - 1/4000. Detects a band of approximately 24.7 kDa (predicted molecular weight: 26.6 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR