Anti-CDNK1A Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX14975
89360-076EA 821.22 CAD
89360-076
Anti-CDNK1A Rabbit Polyclonal Antibody
Antibodies
Cell cycle progression is regulated by cyclins and their cognate Cdks. p21 (WAF1/CIP1) inhibits the activity of each member of the cyclin/Cdk family, and over expression of this protein inhibits the proliferation of mammalian cells. The expression of p21 is inducible by a wide range of stress stimuli. p21 is tightly regulated at the transcriptional level by p53, and probably serves as the effector of p53 cell cycle control.

WB: Use at a dilution of 1/250 - 1/500. Detects a band of approximately 21 kDa (predicted molecular weight: 19.7 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CDNK1A
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR