Anti-P2RX2 Guinea Pig Polyclonal Antibody

Supplier: GeneTex

GTX10262
89349-666EA 624.75 CAD
89349-666
Anti-P2RX2 Guinea Pig Polyclonal Antibody
Antibodies
Guinea pig polyclonal to P2X2

ICC: Use at a dilution of 1/500. IHC-Fr: Use at a dilution of 1/500. WB: Use at a dilution of 1/500. Predicted molecular weight: 47.8 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: P2RX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea Pig
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR