Anti-P2RX2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX10266
89349-680EA 624.75 CAD
89349-680
Anti-P2RX2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to P2X2

ICC: Use at a dilution of 1/1000. IHC-Fr: Use at a dilution of 1/1000. WB: Use at a dilution of 1/1000. Predicted molecular weight: 47,8 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: P2RX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR