Anti-PUMA Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX70455
89364-992EA 565.62 CAD
89364-992
Anti-PUMA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to PUMA

For WB: Use at a dilution of 1:500-1:1,000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: PUMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR