Anti-MELK Goat Polyclonal Antibody

GTX29696
89367-648EA 664.42 CAD
89367-648
Anti-MELK Goat Polyclonal Antibody
Type: Primary
Antigen: MELK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR