Anti-MMP10 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX25708
89361-712EA 717.64 CAD
89361-712
Anti-MMP10 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to MMP10

ELISA: Use at a concentration of 0.1 - 1.0 ?g/ml. IP: Use 3-5?g for 10^6 cells. WB: Use at a concentration of 0.5 - 2.0 ?g/ml. Predicted molecular weight: 54 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MMP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR