Anti-Aflatoxin Rat Monoclonal Antibody

Supplier: GeneTex

GTX20685
89360-876EA 565.91 CAD
89360-876
Anti-Aflatoxin Rat Monoclonal Antibody
Antibodies
Type: Primary
Antigen: [delete]
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR