Anti-AFP Rabbit Monoclonal Antibody

GTX61406
89363-972EA 556.26 CAD
89363-972
Anti-AFP Rabbit Monoclonal Antibody
Alpha-fetoprotein (AFP) is a serum glycoprotein protein produced in the liver or yolk sac of fetal staged mammals. AFP synthesis is minimal after birth and trace amount is expressed in the adult liver . AFP gene expression is regulated by the interactions between steroid hormone receptors and transcriptional factors in separate signal transduction pathways . AFP functions as a binding and transporting ligand and cell growth regulator. Elevated expression level of AFP has been implicated in colorectal, ovarian, pancreatic, testicular, and certain liver cancers. High levels of AFP is also seen in some diseases such as hepatitis and colitis. AFP is used as a screening marker for fetal abnormalities in pregnant women such as Down syndrome. Recently, AFP has been introduced as an anti-cancer drugs-ligand carrier, transporting drugs to target tumor cells, increasing anti-tumor efficiency .

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR