Anti-TUBB Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX48618
89352-892EA 915.69 CAD
89352-892
Anti-TUBB Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to beta Tubulin (Biotin)

Western Blot 1:1000-1:10000

Type: Primary
Antigen: TUBB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR