Anti-TUBB Mouse Monoclonal Antibody [clone: TU-06]

Supplier: GeneTex
GTX27792
89362-668EA 946.55 CAD
89362-668
Anti-TUBB Mouse Monoclonal Antibody [clone: TU-06]
Antibodies
Mouse monoclonal [TU - 06] to beta Tubulin

ELISA: Use at an assay dependant dilution. ICC: Use at an assay dependant dilution. IP: Use at an assay dependant dilution. WB: Use at a dilution of 1/1000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TUBB
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Carp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR